Merck
  • Site-specific fluorescent labeling approaches for naringenin, an essential flavonone in plant nitrogen-fixation signaling pathways.
Site-specific fluorescent labeling approaches for naringenin, an essential flavonone in plant nitrogen-fixation signaling pathways.
The Journal of organic chemistry (2008-10-02)
Lei Chen, Feng-Qing Li, Bi-He Hou, Guo-Fan Hong, Zhu-Jun Yao
ABSTRACT
In search of an appropriate position for the fluorescent labeling, six chemically available positions of the flavonone core of naringenin have been examined. A number of azido-containing naringenin derivatives were accordingly prepared in various site-specific fashions, and the mild Cu(I)-catalyzed Huisgen 1,3-dipolar cycloaddition successfully served as the common "Click" labeling tool in the final steps. On the basis of the biological activities of the first batch of labeled compounds, further optimization at the C-6 position of naringenin finally afforded naringenin-flu (27), which acquired 20% of the potency of naringenin and presented good optical properties. Entry of naringenin-flu into living Rhizobium cells was demonstrated by in vitro fluorescent imaging experiments.