Merck
  • Cyclooxygenase 2 gene polymorphisms and chronic periodontitis in a North Indian population: a pilot study.
Cyclooxygenase 2 gene polymorphisms and chronic periodontitis in a North Indian population: a pilot study.
Journal of periodontal & implant science (2012-11-28)
Anika Daing, Sarvendra Vikram Singh, Charanjeet Singh Saimbi, Mohammad Akhlaq Khan, Srikanta Kumar Rath
ABSTRACT
Cyclooxygenase (COX) enzyme catalyzes the production of prostaglandins, which are important mediators of tissue destruction in periodontitis. Single nucleotide polymorphisms of COX2 enzyme have been associated with increasing susceptibility to inflammatory diseases. The present study evaluates the association of two single nucleotide polymorphisms in COX2 gene (-1195G>A and 8473C>T) with chronic periodontitis in North Indians. Both SNPs and their haplotypes were used to explore the associations between COX2 polymorphisms and chronic periodontitis in 56 patients and 60 controls. Genotyping was done by polymerase chain reaction followed by restriction fragment length polymorphism. Chi-square test and logistic regression analysis were performed for association analysis. By the individual genotype analysis, mutant genotypes (GA and AA) of COX2 -1195 showed more than a two fold risk (odds ratio [OR]>2) and COX2 8473 (TC and CC) showed a reduced risk for the disease, but the findings were not statistically significant. Haplotype analysis showed that the frequency of the haplotype AT was higher in the case group and a significant association was found for haplotype AT (OR, 1.79; 95% confidence interval, 1.03 to 3.11; P=0.0370) indicating an association between the AT haplotype of COX2 gene SNPs and chronic periodontitis. Individual genotypes of both the SNPs were not associated while haplotype AT was found to be associated with chronic periodontitis in North Indians.
Sigma-Aldrich
GenElute Blood Genomic DNA Kit, sufficient for 350 purifications
Pricing and availability is not currently available.
Sigma-Aldrich
GenElute Blood Genomic DNA Kit, sufficient for 70 purifications
Pricing and availability is not currently available.