Merck
  • VEGF expression disparities in brainstem motor neurons of the SOD1G93A ALS model: Correlations with neuronal vulnerability.
VEGF expression disparities in brainstem motor neurons of the SOD1G93A ALS model: Correlations with neuronal vulnerability.
Neurotherapeutics : the journal of the American Society for Experimental NeuroTherapeutics (2024-03-13)
Silvia Silva-Hucha, M Estrella Fernández de Sevilla, Kirsty M Humphreys, Fiona E Benson, Jaime M Franco, David Pozo, Angel M Pastor, Sara Morcuende
ABSTRACT
Amyotrophic lateral sclerosis (ALS) is a rare neuromuscular disease characterized by severe muscle weakness mainly due to degeneration and death of motor neurons. A peculiarity of the neurodegenerative processes is the variable susceptibility among distinct neuronal populations, exemplified by the contrasting resilience of motor neurons innervating the ocular motor system and the more vulnerable facial and hypoglossal motor neurons. The crucial role of vascular endothelial growth factor (VEGF) as a neuroprotective factor in the nervous system is well-established since a deficit of VEGF has been related to motoneuronal degeneration. In this study, we investigated the survival of ocular, facial, and hypoglossal motor neurons utilizing the murine SOD1G93A ALS model at various stages of the disease. Our primary objective was to determine whether the survival of the different brainstem motor neurons was linked to disparate VEGF expression levels in resilient and susceptible motor neurons throughout neurodegeneration. Our findings revealed a selective loss of motor neurons exclusively within the vulnerable nuclei. Furthermore, a significantly higher level of VEGF was detected in the more resistant motor neurons, the extraocular ones. We also examined whether TDP-43 dynamics in the brainstem motor neuron of SOD mice was altered. Our data suggests that the increased VEGF levels observed in extraocular motor neurons may potentially underlie their resistance during the neurodegenerative processes in ALS in a TDP-43-independent manner. Our work might help to better understand the underlying mechanisms of selective vulnerability of motor neurons in ALS.
Sigma-Aldrich
Anti-Glyceraldehyde-3-Phosphate Dehydrogenase Antibody, clone 6C5, clone 6C5, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-TDP-43 Antibody, clone DB9, clone DB9, from mouse
Pricing and availability is not currently available.
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Protease Inhibitor Cocktail Tablets provided in a glass vial, Tablets provided in a glass vial
Pricing and availability is not currently available.