Merck
  • Erythrocyte scaffolding protein p55/MPP1 functions as an essential regulator of neutrophil polarity.
Erythrocyte scaffolding protein p55/MPP1 functions as an essential regulator of neutrophil polarity.
Proceedings of the National Academy of Sciences of the United States of America (2009-11-10)
Brendan J Quinn, Emily J Welch, Anthony C Kim, Mary A Lokuta, Anna Huttenlocher, Anwar A Khan, Shafi M Kuchay, Athar H Chishti
ABSTRACT
As mediators of innate immunity, neutrophils respond to chemoattractants by adopting a highly polarized morphology. Efficient chemotaxis requires the formation of one prominent pseudopod at the cell front characterized by actin polymerization, while local inhibition suppresses the formation of rear and lateral protrusions. This asymmetric control of signaling pathways is required for directional migration along a chemotactic gradient. Here, we identify the MAGUK protein p55/MPP1 as a mediator of the frontness signal required for neutrophil polarization. We developed a p55 knockout (p55(-/-)) mouse model, and demonstrate that p55(-/-) neutrophils form multiple transient pseudopods upon chemotactic stimulation, and do not migrate efficiently in vitro. Upon agonist stimulation, p55 is rapidly recruited to the leading edge of neutrophils in mice and humans. Total F-actin polymerization, along with Rac1 and RhoA activation, appear to be normal in p55(-/-) neutrophils. Importantly, phosphorylation of Akt is significantly decreased in p55(-/-) neutrophils upon chemotactic stimulation. The activity of immunoprecipitated phosphatidylinositol 3-kinase gamma (PI3Kgamma), responsible for chemoattractant-induced synthesis of PIP(3) and Akt phosphorylation, is unperturbed in p55(-/-) neutrophils. Although the total amount of PIP(3) is normal in p55(-/-) neutrophils, PIP(3) is diffusely localized and forms punctate aggregates in activated p55(-/-) neutrophils, as compared to its accumulation at the leading edge membrane in the wild type neutrophils. Together, these results show that p55 is required for neutrophil polarization by regulating Akt phosphorylation through a mechanism that is independent of PI3Kgamma activity.
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (25 °C), suitable for cell culture
Sigma-Aldrich
Percoll®, suitable for plant cell culture
Sigma-Aldrich
Percoll®, pH 8.5-9.5 (20 °C)