Merck
  • Calcitriol Treatment Is Safe and Increases Frataxin Levels in Friedreich Ataxia Patients.
Calcitriol Treatment Is Safe and Increases Frataxin Levels in Friedreich Ataxia Patients.
Movement disorders : official journal of the Movement Disorder Society (2024-05-02)
Berta Alemany-Perna, Jordi Tamarit, Elisa Cabiscol, Fabien Delaspre, Albert Miguela, Joana Maria Huertas-Pons, Ana Quiroga-Varela, Miguel Merchan Ruiz, Daniel López Domínguez, Lluís Ramió I Torrentà, David Genís, Joaquim Ros
ABSTRACT
Calcitriol, the active form of vitamin D (also known as 1,25-dihydroxycholecalciferol), improves the phenotype and increases frataxin levels in cell models of Friedreich ataxia (FRDA). Based on these results, we aimed measuring the effects of a calcitriol dose of 0.25 mcg/24h in the neurological function and frataxin levels when administered to FRDA patients for a year. 20 FRDA patients where recluted and 15 patients completed the treatment for a year. Evaluations of neurological function changes (SARA scale, 9-HPT, 8-MWT, PATA test) and quality of life (Barthel Scale and Short Form (36) Health Survey [SF-36] quality of life questionnaire) were performed. Frataxin amounts were measured in isolated platelets obtained from these FRDA patients, from heterozygous FRDA carriers (relatives of the FA patients) and from non-heterozygous sex and age matched controls. Although the patients did not experience any observable neurological improvement, there was a statistically significant increase in frataxin levels from initial values, 5.5 to 7.0 pg/μg after 12 months. Differences in frataxin levels referred to total protein levels were observed among sex- and age-matched controls (18.1 pg/μg), relative controls (10.1 pg/μg), and FRDA patients (5.7 pg/μg). The treatment was well tolerated by most patients, and only some of them experienced minor adverse effects at the beginning of the trial. Calcitriol dosage used (0.25 mcg/24 h) is safe for FRDA patients, and it increases frataxin levels. We cannot rule out that higher doses administered longer could yield neurological benefits. © 2024 The Authors. Movement Disorders published by Wiley Periodicals LLC on behalf of International Parkinson and Movement Disorder Society.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Anti-Rabbit IgG (H+L), F(ab′)2 fragment, CF640R antibody produced in goat, ~2 mg/mL, affinity isolated antibody, buffered aqueous solution