Merck
  • Heat-activated nanomedicine formulation improves the anticancer potential of the HSP90 inhibitor luminespib in vitro.
Heat-activated nanomedicine formulation improves the anticancer potential of the HSP90 inhibitor luminespib in vitro.
Scientific reports (2021-05-29)
Brittany Epp-Ducharme, Michael Dunne, Linyu Fan, James C Evans, Lubabah Ahmed, Pauric Bannigan, Christine Allen
ABSTRACT
The heat shock protein 90 inhibitor, luminespib, has demonstrated potent preclinical activity against numerous cancers. However, clinical translation has been impeded by dose-limiting toxicities that have necessitated dosing schedules which have reduced therapeutic efficacy. As such, luminespib is a prime candidate for reformulation using advanced drug delivery strategies that improve tumor delivery efficiency and limit off-target side effects. Specifically, thermosensitive liposomes are proposed as a drug delivery strategy capable of delivering high concentrations of drug to the tumor in combination with other chemotherapeutic molecules. Indeed, this work establishes that luminespib exhibits synergistic activity in lung cancer in combination with standard of care drugs such as cisplatin and vinorelbine. While our research team has previously developed thermosensitive liposomes containing cisplatin or vinorelbine, this work presents the first liposomal formulation of luminespib. The physico-chemical properties and heat-triggered release of the formulation were characterized. Cytotoxicity assays were used to determine the optimal drug ratios for treatment of luminespib in combination with cisplatin or vinorelbine in non-small cell lung cancer cells. The formulation and drug combination work presented in this paper offer the potential for resuscitation of the clinical prospects of a promising anticancer agent.
Sigma-Aldrich
NVP-AUY922, ≥98% (HPLC)