Merck
  • Characterization of serotonin-5-HTR1E signaling pathways and its role in cell survival.
Characterization of serotonin-5-HTR1E signaling pathways and its role in cell survival.
Research square (2023-02-16)
Vinay Kumar Sharma, Kiersten Campbell, Xuyu Yang, Ryan Dale, Y Peng Loh
ABSTRACT
5-Hydroxy tryptamine receptor 1E (5-HTR1E) is reported to activate cAMP and ERK pathways via its ligands and binding partners, but the detailed mechanism underlying the serotonin induced 5-HTR1E signaling is still not known. In the present study, we determined the cellular regulators of ERK and cAMP signaling pathways in response to serotonin induced 5-HTR1E activation in 5-HTR1E overexpressing HEK293 cells. We found that Pertussis Toxin (PTX) treatment completely reversed the effect of serotonin-5-HTR1E mediated signaling on cAMP and ERK pathways, confirming the involvement of a Gαi-linked cascade. We also observed that Gβγ and Gq were not associated with 5-HTR1E activation, while blocking PKA inhibited ERK signaling only, and had no effect on cAMP. Additionally, serotonin-stimulated ERK1/2 phosphorylation was similar in 5-HTR1E overexpressing, β-arrestin-deficient HEK293 cells and is solely dependent on G protein signaling. siRNA mediated gene knockout studies in SH-SY5Y cells revealed that the inhibition of 5-HTR1E reduced the expression of cMyc, Cyclin D1, Cyclin E and BCL2 genes which are related to cell cycle regulation and survival. MTT assays showed that 5-HTR1E knockdown in SHSY-5Y and U118 cells inhibited cell survival significantly. In addition to the signaling mechanism, we also performed RNA-seq analysis in 5-HTR1E overexpressing HEK293 cells and found that 5-HTR1E can regulate the expression of Receptor activity modifying protein 1 ( RAMP1 ), Nuclear receptor 1 ( NR4A1 ) and other Cyclin genes. These findings indicate that serotonin interaction with 5-HTR1E receptor simultaneously activates cAMP and ERK pathway in HEK293 cells and its expression is important for cell survival.