Merck
Oligonucleotide Bioconjugation with Bifunctional Palladium Reagents.
Angewandte Chemie (International ed. in English) (2021-03-18)
Muhammad Jbara, Jacob Rodriguez, Heemal H Dhanjee, Andrei Loas, Stephen L Buchwald, Bradley L Pentelute
ABSTRACT
Organometallic reagents enable practical strategies for bioconjugation. Innovations in the design of water-soluble ligands and the enhancement of reaction rates have allowed for chemoselective cross-coupling reactions of peptides and proteins to be carried out in water. There are currently no organometallic-based methods for oligonucleotide bioconjugation to other biomolecules. Here we report bifunctional palladium(II)-oxidative addition complexes (OACs) as reagents for high-yielding oligonucleotide bioconjugation reactions. These bifunctional OACs react chemoselectively with amine-modified oligonucleotides to generate the first isolable, bench stable oligonucleotide-palladium(II) OACs. These complexes undergo site-selective C-S arylation with a broad range of native thiol-containing biomolecules at low micromolar concentrations in under one hour. This approach provided oligonucleotide-peptide, oligonucleotide-protein, oligonucleotide-small molecule, and oligonucleotide-oligonucleotide conjugates in >80 % yield and afforded conjugation of multiple copies of oligonucleotides onto a monoclonal antibody.
Sigma-Aldrich
SPhos Pd G6 acylation, ≥95%
Pricing and availability is not currently available.