Merck
  • FAM69C functions as a kinase for eIF2α and promotes stress granule assembly.
FAM69C functions as a kinase for eIF2α and promotes stress granule assembly.
EMBO reports (2023-03-18)
Zhongyan Wu, Fan Mei, Yangyang Gan, Anhang Liu, Jiapan Hu, Yan Jin, Yuxin Yin
ABSTRACT
Stress granules are dynamic cytoplasmic ribonucleoprotein granules that assemble in response to cellular stress. Aberrant formation of stress granules has been linked to neurodegenerative diseases. However, the molecular mechanisms underlying the initiation of stress granules remain elusive. Here we report that the brain-enriched protein kinase FAM69C promotes stress granule assembly through phosphorylation of eukaryotic translation initiation factor 2 (eIF2α). FAM69C physically interacts with eIF2α and functions as a stress-specific kinase for eIF2α, leading to stress-induced protein translation arrest and stress granule assembly. Primary microglia derived from Fam69c knockout mice exhibit aberrant stress granule assembly in response to oxidative stress and ATP. Defective stress granule assembly in microglia correlates with the formation of ASC specks and NLRP3 inflammasome activation, whereas induction of stress granule precludes inflammasome formation. Consistently, increased NLRP3 levels, caspase-1 cleavage and Il18 expression corroborate microglia-associated neuroinflammation in aged Fam69c knockout mice. Our study demonstrates that FAM69C is critical for stress granule assembly and suggests its role in the regulation of microglia function.
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Puromycin Antibody, clone 17H1, clone 17H1, from rat
Pricing and availability is not currently available.