Merck
  • Dynamic quality control machinery that operates across compartmental borders mediates the degradation of mammalian nuclear membrane proteins.
Dynamic quality control machinery that operates across compartmental borders mediates the degradation of mammalian nuclear membrane proteins.
Cell reports (2022-11-24)
Pei-Ling Tsai, Christopher J F Cameron, Maria Fernanda Forni, Renee R Wasko, Brigitte S Naughton, Valerie Horsley, Mark B Gerstein, Christian Schlieker
ABSTRACT
Many human diseases are caused by mutations in nuclear envelope (NE) proteins. How protein homeostasis and disease etiology are interconnected at the NE is poorly understood. Specifically, the identity of local ubiquitin ligases that facilitate ubiquitin-proteasome-dependent NE protein turnover is presently unknown. Here, we employ a short-lived, Lamin B receptor disease variant as a model substrate in a genetic screen to uncover key elements of NE protein turnover. We identify the ubiquitin-conjugating enzymes (E2s) Ube2G2 and Ube2D3, the membrane-resident ubiquitin ligases (E3s) RNF5 and HRD1, and the poorly understood protein TMEM33. RNF5, but not HRD1, requires TMEM33 both for efficient biosynthesis and function. Once synthesized, RNF5 responds dynamically to increased substrate levels at the NE by departing from the endoplasmic reticulum, where HRD1 remains confined. Thus, mammalian protein quality control machinery partitions between distinct cellular compartments to address locally changing substrate loads, establishing a robust cellular quality control system.
Product Description
Roche
Anti-HA-Peroxidase, High Affinity, from rat IgG1
Roche
Anti-HA Affinity Matrix, from rat IgG1
Sigma-Aldrich
Anti-RNF5 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)