Merck
  • Natural product P57 induces hypothermia through targeting pyridoxal kinase.
Natural product P57 induces hypothermia through targeting pyridoxal kinase.
Nature communications (2023-09-27)
Ruina Wang, Lei Xiao, Jianbo Pan, Guangsen Bao, Yunmei Zhu, Di Zhu, Jun Wang, Chengfeng Pei, Qinfeng Ma, Xian Fu, Ziruoyu Wang, Mengdi Zhu, Guoxiang Wang, Ling Gong, Qiuping Tong, Min Jiang, Junchi Hu, Miao He, Yun Wang, Tiejun Li, Chunmin Liang, Wei Li, Chunmei Xia, Zengxia Li, Dengke K Ma, Minjia Tan, Jun Yan Liu, Wei Jiang, Cheng Luo, Biao Yu, Yongjun Dang
ABSTRACT
Induction of hypothermia during hibernation/torpor enables certain mammals to survive under extreme environmental conditions. However, pharmacological induction of hypothermia in most mammals remains a huge challenge. Here we show that a natural product P57 promptly induces hypothermia and decreases energy expenditure in mice. Mechanistically, P57 inhibits the kinase activity of pyridoxal kinase (PDXK), a key metabolic enzyme of vitamin B6 catalyzing phosphorylation of pyridoxal (PL), resulting in the accumulation of PL in hypothalamus to cause hypothermia. The hypothermia induced by P57 is significantly blunted in the mice with knockout of PDXK in the preoptic area (POA) of hypothalamus. We further found that P57 and PL have consistent effects on gene expression regulation in hypothalamus, and they may activate medial preoptic area (MPA) neurons in POA to induce hypothermia. Taken together, our findings demonstrate that P57 has a potential application in therapeutic hypothermia through regulation of vitamin B6 metabolism and PDXK serves as a previously unknown target of P57 in thermoregulation. In addition, P57 may serve as a chemical probe for exploring the neuron circuitry related to hypothermia state in mice.
Sigma-Aldrich
Ethacrynic acid, ≥97% (HPLC)
Sigma-Aldrich
Celastrol, ≥98% (HPLC), solid