Merck
  • Rac1, the actin cytoskeleton and microtubules are key players in clathrin-independent endophilin-A3-mediated endocytosis.
Rac1, the actin cytoskeleton and microtubules are key players in clathrin-independent endophilin-A3-mediated endocytosis.
Journal of cell science (2022-06-16)
François Tyckaert, Natacha Zanin, Pierre Morsomme, Henri-François Renard
ABSTRACT
Endocytic mechanisms actively regulate plasma membrane composition and sustain fundamental cellular functions. Recently, we identified a clathrin-independent endocytic (CIE) modality mediated by the BAR domain protein endophilin-A3 (endoA3, encoded by SH3GL3), which controls the cell surface homeostasis of the tumor marker CD166 (also known as ALCAM). Deciphering the molecular machinery of endoA3-dependent CIE should therefore contribute to a better understanding of its pathophysiological role, which remains so far unknown. Here, we investigate the role of actin, Rho GTPases and microtubules, which are major players in CIE processes, in this mechanism. We show that the actin cytoskeleton is dynamically associated with endoA3- and CD166-positive endocytic carriers, and that its perturbation strongly inhibits the process of CD166 uptake. We also reveal that the Rho GTPase Rac1, but not Cdc42, is a master regulator of this endocytic route. Finally, we provide evidence that microtubules and kinesin molecular motors are required to potentiate endoA3-dependent endocytosis. Of note, our study also highlights potential compensation phenomena between endoA3-dependent CIE and macropinocytosis. Altogether, our data deepen our understanding of this CIE modality and further differentiate it from other unconventional endocytic mechanisms. This article has an associated First Person interview with the first author of the paper.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
ML 141, ≥98% (HPLC)
Sigma-Aldrich
Anti-Myosin IIA, non muscle antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, ascites fluid, clone B-5-1-2
Sigma-Aldrich
Monastrol, ≥98% (HPLC), solid