Merck
  • The circadian clock protein Cryptochrome 1 is a direct target and feedback regulator of the Hippo pathway.
The circadian clock protein Cryptochrome 1 is a direct target and feedback regulator of the Hippo pathway.
iScience (2023-08-18)
Abdelhalim Azzi, Zhipeng Tao, Yang Sun, Hannah Erb, Carla Guarino, Xu Wu
ABSTRACT
Circadian clock controls daily behavior and physiology. The activity of various signaling pathways affects clock gene expression. Here, we show that the core circadian clock gene CRY1 is a direct target of the Hippo pathway effector YAP. YAP binds to TEADs and occupies the proximal promoter regions of CRY1, positively regulating its transcription. Interestingly, we further identified that CRY1 acts in a feedback loop to fine-tune Hippo pathway activation by modulating the expression of YAP and MOB1. Indeed, loss of CRY1 results in enhanced YAP activation. Consistently, we found that YAP levels and activity control clock gene expression and oscillation in synchronized cells. Furthermore, in breast cancer cells, CRY1 downregulation causes YAP/TAZ hyperactivation and enhanced DNA damage. Together, our findings provide a direct mechanistic link between the Hippo pathway and the circadian clock, where CRY1 and Hippo components form an orchestrated signaling network that influences cell growth and circadian rhythm.
Product Description
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Pricing and availability is not currently available.
Sigma-Aldrich
KL001, ≥98% (HPLC)
Pricing and availability is not currently available.