Merck
  • Mapping Lysine Acetyltransferase-Ligand Interactions by Activity-Based Capture.
Mapping Lysine Acetyltransferase-Ligand Interactions by Activity-Based Capture.
Methods in enzymology (2016-07-18)
D C Montgomery, J L Meier
ABSTRACT
Changes in reversible protein acetylation mediate many key aspects of genomic regulation and enzyme function. The catalysts for this posttranslational modification, lysine acetyltransferases (KATs), have been difficult targets for characterization due to their complex architecture and challenging reconstitution. To address this challenge, here we describe methods to profile endogenous KAT activities using activity-based probes. This method facilitates the targeted analysis of several cellular KATs and can be used to study their interactions with many different types of ligands, including acyl-CoA metabolites. This competitive activity-based capture approach provides a method to assess the selectivity of ligands for different KAT families in complex proteomic settings, and thus has the potential to offer substantial insights into the regulation of cellular KAT function.
Sigma-Aldrich
N-(3-Azidopropyl)biotinamide, ≥95%