Merck
  • The effect of Dnaaf5 gene dosage on primary ciliary dyskinesia phenotypes.
The effect of Dnaaf5 gene dosage on primary ciliary dyskinesia phenotypes.
JCI insight (2023-04-27)
Amjad Horani, Deepesh Kumar Gupta, Jian Xu, Huihui Xu, Lis Del Carmen Puga-Molina, Celia M Santi, Sruthi Ramagiri, Steven K Brennan, Jiehong Pan, Jeffrey R Koenitzer, Tao Huang, Rachael M Hyland, Sean P Gunsten, Shin-Cheng Tzeng, Jennifer M Strahle, Pleasantine Mill, Moe R Mahjoub, Susan K Dutcher, Steven L Brody
ABSTRACT
DNAAF5 is a dynein motor assembly factor associated with the autosomal heterogenic recessive condition of motile cilia, primary ciliary dyskinesia (PCD). The effects of allele heterozygosity on motile cilia function are unknown. We used CRISPR-Cas9 genome editing in mice to recreate a human missense variant identified in patients with mild PCD and a second, frameshift-null deletion in Dnaaf5. Litters with Dnaaf5 heteroallelic variants showed distinct missense and null gene dosage effects. Homozygosity for the null Dnaaf5 alleles was embryonic lethal. Compound heterozygous animals with the missense and null alleles showed severe disease manifesting as hydrocephalus and early lethality. However, animals homozygous for the missense mutation had improved survival, with partially preserved cilia function and motor assembly observed by ultrastructure analysis. Notably, the same variant alleles exhibited divergent cilia function across different multiciliated tissues. Proteomic analysis of isolated airway cilia from mutant mice revealed reduction in some axonemal regulatory and structural proteins not previously reported in DNAAF5 variants. Transcriptional analysis of mouse and human mutant cells showed increased expression of genes coding for axonemal proteins. These findings suggest allele-specific and tissue-specific molecular requirements for cilia motor assembly that may affect disease phenotypes and clinical trajectory in motile ciliopathies.
Sigma-Aldrich
Anti-CFAP126 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-NME9 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.