Merck
  • An HDAC6 inhibitor reverses chemotherapy-induced mechanical hypersensitivity via an IL-10 and macrophage dependent pathway.
An HDAC6 inhibitor reverses chemotherapy-induced mechanical hypersensitivity via an IL-10 and macrophage dependent pathway.
Brain, behavior, and immunity (2021-12-17)
Jixiang Zhang, Jiacheng Ma, Ronnie T Trinh, Cobi J Heijnen, Annemieke Kavelaars
ABSTRACT
Chemotherapy-induced peripheral neuropathy (CIPN) impacts a growing number of cancer survivors and treatment options are limited. Histone deacetylase 6 (HDAC6) inhibitors are attractive candidates because they reverse established CIPN and may enhance anti-tumor effects of chemotherapy. Before considering clinical application of HDAC6 inhibitors, the mechanisms underlying reversal of CIPN need to be identified. We showed previously that deletion of Hdac6 from sensory neurons did not prevent cisplatin-induced mechanical hypersensitivity, while global deletion of Hdac6 was protective, indicating involvement of HDAC6 in other cell types. Here we show that local depletion of MRC1 (CD206)-positive macrophages without affecting microglia by intrathecal administration of mannosylated clodronate liposomes reduced the capacity of an HDAC6 inhibitor to reverse cisplatin-induced mechanical hypersensitivity. The HDAC6 inhibitor increased spinal cord Il10 mRNA and this was M2-macrophage dependent. Intrathecal administration of anti-IL-10 antibody or genetic deletion of Il10 prevented resolution of mechanical hypersensitivity. Genetic deletion of the IL-10 receptor from Advillin+ neurons prevented resolution of mechanical hypersensitivity in mice treated with the HDAC6 inhibitor. These findings indicate that treatment with an HDAC6 inhibitor increases macrophage-derived IL-10 signaling to IL-10 receptors on Advillin+ sensory neurons to resolve mechanical hypersensitivity. Cisplatin decreases mitochondrial function in sensory axons, and HDAC6 inhibition can promote axonal transport of healthy mitochondria. Indeed, the HDAC6 inhibitor normalized cisplatin-induced tibial nerve mitochondrial deficits. However, this was independent of macrophages and IL-10 signaling. In conclusion, our findings indicate that administration of an HDAC6 inhibitor reverses cisplatin-induced mechanical hypersensitivity through two complementary pathways: macrophage HDAC6 inhibition to promote IL-10 production and IL-10 signaling to DRG neurons, and neuronal HDAC6 inhibition to restore axonal mitochondrial health.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Rotenone, ≥95%
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
IgG from goat serum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Anti-Interleukin-10 antibody produced in goat, IgG fraction of antiserum, lyophilized powder