Merck
  • High-resolution microbial community reconstruction by integrating short reads from multiple 16S rRNA regions.
High-resolution microbial community reconstruction by integrating short reads from multiple 16S rRNA regions.
Nucleic acids research (2013-11-12)
Amnon Amir, Amit Zeisel, Or Zuk, Michael Elgart, Shay Stern, Ohad Shamir, Peter J Turnbaugh, Yoav Soen, Noam Shental
ABSTRACT
The emergence of massively parallel sequencing technology has revolutionized microbial profiling, allowing the unprecedented comparison of microbial diversity across time and space in a wide range of host-associated and environmental ecosystems. Although the high-throughput nature of such methods enables the detection of low-frequency bacteria, these advances come at the cost of sequencing read length, limiting the phylogenetic resolution possible by current methods. Here, we present a generic approach for integrating short reads from large genomic regions, thus enabling phylogenetic resolution far exceeding current methods. The approach is based on a mapping to a statistical model that is later solved as a constrained optimization problem. We demonstrate the utility of this method by analyzing human saliva and Drosophila samples, using Illumina single-end sequencing of a 750 bp amplicon of the 16S rRNA gene. Phylogenetic resolution is significantly extended while reducing the number of falsely detected bacteria, as compared with standard single-region Roche 454 Pyrosequencing. Our approach can be seamlessly applied to simultaneous sequencing of multiple genes providing a higher resolution view of the composition and activity of complex microbial communities.
Sigma-Aldrich
5R-Plex kit, Ultra-sensitive 16S NGS assay for degraded and low biomass DNA
Pricing and availability is not currently available.