Merck
  • Multiple phosphorylation of chicken protein tyrosine phosphatase 1 and human protein tyrosine phosphatase 1B by casein kinase II and p60c-src in vitro.
Multiple phosphorylation of chicken protein tyrosine phosphatase 1 and human protein tyrosine phosphatase 1B by casein kinase II and p60c-src in vitro.
Biochemical and biophysical research communications (1998-05-26)
E J Jung, Y S Kang, C W Kim
ABSTRACT
We have cloned a soluble chicken protein tyrosine phosphatase, named CPTP1, from the cDNA library of chicken intestine. The CPTP1 showed 92% sequence identity to the corresponding 321 amino acid residues of human PTP1B (HPTP1B). CPTP1 lacked 13 amino acids of the N-terminal region compared with HPTP1B, while the C-terminal 48 amino acid sequence of this protein was distinct from those of other PTPs. In vitro phosphorylation and phosphoamino acid analysis showed that both CPTP1 and HPTP1B were phosphorylated on serine and threonine residues near their N-terminus by casein kinase II (CKII). Furthermore, phosphorylation of CPTP1 by CKII resulted in an inhibition of its phosphatase activity in vitro. Interestingly, both CPTP1 and HPTP1B were also tyrosine-phosphorylated near their N-terminus by p60c-src. When we examined the vanadate effect, in the absence of vanadate, the tyrosine-phosphorylated CPTP1 by p60c-src was autodephosphorylated by its own phosphatase activity. These results suggest that both CPTP1 and HPTP1B might play an important role in CKII- and p60c-src-induced signal transduction cascades.
Sigma-Aldrich
PTP1B full length Active human, recombinant, expressed in E. coli, ≥55% (SDS-PAGE), aqueous solution
Pricing and availability is not currently available.