Merck
  • Decitabine enhances the tumoricidal potential of TRAIL via the epigenetic regulation of death receptor 4 in gastric cancer.
Decitabine enhances the tumoricidal potential of TRAIL via the epigenetic regulation of death receptor 4 in gastric cancer.
Journal of gastrointestinal oncology (2023-01-14)
Lin Li, Qian Niu, Yuanmin Zhu, Biao Fan, Guibin Yang
ABSTRACT
Deoxyribonucleic acid (DNA) methyltransferase inhibitors, such as decitabine, have made great advances in cancer therapy as combinational drugs. Tumor necrosis factor-related apoptosis-inducing ligand (TRAIL) has an obvious anti-tumor effect; however, some gastric cancer (GC) cells are resistant to TRAIL-induced cell death. This study sought to explore the synergistic anti-tumor effect of TRAIL and decitabine, and the potential synergetic mechanism. The cell growth inhibition effect was monitored by the IncuCyte ZOOM Live-Cell Analysis System, and cell viability was determined by Cell Counting Kit-8 assays. Apoptosis was detected by Annexin V/Propidium Iodide double staining. Death receptor 4 (DR4) was knocked down by ribonucleic acid (RNA) interference, and the effect of DR4 deletion on TRAIL sensitivity was analyzed. Methylation-specific polymerase chain reaction (PCR) was applied to determine the methylation status of DR4. The messenger RNA (mRNA) and protein expression levels were detected by quantitative real-time polymerase chain reaction (qRT-PCR) and western blot. The expression of the DRs on the cell membrane surfaces was analyzed by flow cytometry. The combined use of decitabine and TRAIL synergistically inhibited cell growth in 2 TRAIL-resistant cell lines. Further, decitabine augmented TRAIL-induced apoptosis in a caspase-dependent manner. The co-application of decitabine and TRAIL facilitated the activation of caspase-7, -8, -9, and poly ADP-ribose polymerase (PARP). Notably, decitabine increased the expression of DR4 at the transcriptional and post-transcriptional levels. DR4 expression on the cell membrane surfaces was also upregulated after decitabine exposure. The depletion of DR4 by specific inhibitors attenuated TRAIL-induced apoptosis and weakened the synergistic effects of decitabine and TRAIL. In addition, DR4 gene presented methylation status in SNU-1 cells. The low mRNA and protein expression of DR4 were also detected in SNU-1 cells. Decitabine enhances the effect of TRAIL by inhibiting the growth and inducing the apoptosis of GC cells. This is achieved by the epigenetic modification of decitabine, which upregulates DR4. Decitabine may act as a sensitizing agent of TRAIL. The combined use of decitabine and TRAIL may provide a novel idea for GC treatment.
Sigma-Aldrich
Anti-TNFRSF10B antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.