Merck
Increased HRD score in cisplatin resistant penile cancer cells.
BMC cancer (2022-12-24)
Ria Winkelmann, Katrin Bankov, Claudia Döring, Jaroslav Cinatl, Sebastian Grothe, Florian Rothweiler, Martin Michaelis, Christina Schmitt, Peter J Wild, Melanie Demes, Jindrich Cinatl, Stefan Vallo
ABSTRACT
Penile cancer is a rare disease in demand for new therapeutic options. Frequently used combination chemotherapy with 5 fluorouracil (5-FU) and cisplatin (CDDP) in patients with metastatic penile cancer mostly results in the development of acquired drug resistance. Availability of cell culture models with acquired resistance against standard therapy could help to understand molecular mechanisms underlying chemotherapy resistance and to identify candidate treatments for an efficient second line therapy. We generated a cell line from a humanpapilloma virus (HPV) negative penile squamous cell carcinoma (UKF-PEC-1). This cell line was subject to chronic exposure to chemotherapy with CDDP and / or 5-FU to induce acquired resistance in the newly established chemo-resistant sublines (PEC-1rCDDP2500, adapted to 2500 ng/ml CDDP; UKF-PEC-1r5-FU500, adapted to 500 ng/ml 5- FU; UKF-PEC1rCDDP2500/r5-FU500, adapted to 2500 ng/ml CDDP and 500 ng/ml 5 -FU). Afterwards cell line pellets were formalin-fixed, paraffin embedded and subject to sequencing as well as testing for homologous recombination deficiency (HRD). Additionally, exemplary immunohistochemical stainings for p53 and gammaH2AX were applied for verification purposes. Finally, UKF-PEC-1rCDDP2500, UKF-PEC-1r5-FU500, UKF-PEC1rCDDP2500/r5-FU500, and UKF-PEC-3 (an alternative penis cancer cell line) were tested for sensitivity to paclitaxel, docetaxel, olaparib, and rucaparib. The chemo-resistant sublines differed in their mutational landscapes. UKF-PEC-1rCDDP2500 was characterized by an increased HRD score, which is supposed to be associated with increased PARP inhibitor and immune checkpoint inhibitor sensitivity in cancer. However, UKF-PEC-1rCDDP2500 did not display sensitivity to PARP inhibitors.
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone 6L16, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells
Pricing and availability is not currently available.