Merck
  • CD-NTase family member MB21D2 promotes cGAS-mediated antiviral and antitumor immunity.
CD-NTase family member MB21D2 promotes cGAS-mediated antiviral and antitumor immunity.
Cell death and differentiation (2023-01-22)
Hansen Liu, Zhenzhen Yan, Deyu Zhu, Haiyan Xu, Feng Liu, Tian Chen, Honghai Zhang, Yi Zheng, Bingyu Liu, Lei Zhang, Wei Zhao, Chengjiang Gao
ABSTRACT
cGAS/DncV-like nucleotidyltransferase (CD-NTase) family members are immune sensors that synthesize diverse nucleotide signals to initiate antiviral response in bacteria and animals. As a founding member of CD-NTase enzyme, cGAS has been identified as a key sensor for cytoplasmic DNA and type I interferons (IFNs) signaling in metazoan. However, the functions of other metazoan CD-NTases remain enigmatic. Here, we showed that Mab-21 domain-containing protein 2 (MB21D2), another member of the CD-NTase family, plays a positive role in modulating the cGAS-STING signaling in myeloid cells. Deficiency of MB21D2 in THP-1 cells or mice macrophages led to impaired production of type I interferon upon DNA stimulation. Consistently, Mb21d2-/- mice showed more susceptible to infection with DNA virus and faster growth of melanoma, compared to its counterparts. Mechanistically, MB21D2 specially bound with the N-terminal of cGAS, facilitated its liquid phase condensation and DNA-binding activity, leading to the enhanced production of cGAMP and subsequent IFN-β production. Thus, our findings unveiled that the CD-NTase family member MB21D2 contributes to host antiviral and antitumor responses by enhancing cGAS activation.
Sigma-Aldrich
Monoclonal Anti-VSV Glycoprotein antibody produced in mouse, clone P5D4, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-C3orf59 (C-terminal) antibody produced in rabbit, affinity isolated antibody
Pricing and availability is not currently available.