- 1,2,6-Thiadiazinones as Novel Narrow Spectrum Calcium/Calmodulin-Dependent Protein Kinase Kinase 2 (CaMKK2) Inhibitors.
1,2,6-Thiadiazinones as Novel Narrow Spectrum Calcium/Calmodulin-Dependent Protein Kinase Kinase 2 (CaMKK2) Inhibitors.
Molecules (Basel, Switzerland) (2018-05-23)
Christopher R M Asquith, Paulo H Godoi, Rafael M Couñago, Tuomo Laitinen, John W Scott, Christopher G Langendorf, Jonathan S Oakhill, David H Drewry, William J Zuercher, Panayiotis A Koutentis, Timothy M Willson, Andreas S Kalogirou
PMID29783765
ABSTRACT
We demonstrate for the first time that 4H-1,2,6-thiadiazin-4-one (TDZ) can function as a chemotype for the design of ATP-competitive kinase inhibitors. Using insights from a co-crystal structure of a 3,5-bis(arylamino)-4H-1,2,6-thiadiazin-4-one bound to calcium/calmodulin-dependent protein kinase kinase 2 (CaMKK2), several analogues were identified with micromolar activity through targeted displacement of bound water molecules in the active site. Since the TDZ analogues showed reduced promiscuity compared to their 2,4-dianilinopyrimidine counter parts, they represent starting points for development of highly selective kinase inhibitors.
Product Description
Millipore
Protease Inhibitor Cocktail Set VII, Protease Inhibitor Cocktail Set VII contains five protease inhibitors with broad specificity. Recommended for purification of proteins containing His•Tag sequences.