Merck
Expression and Activity of Calpain A in Drosophila melanogaster.
Methods in molecular biology (Clifton, N.J.) (2019-01-09)
Maira Cardoso, Danielle Oliveira, Helena Araujo
ABSTRACT
Detecting calpain activity in Drosophila tissues is a fundamental tool to study calpain function. We use differential centrifugation to prepare membrane- versus cytosol-enriched fractions for measuring calpain activity with the fluorogenic substrate N-LY-AMC. With this method one can measure calpain A activity in wild-type flies and in several mutant fly backgrounds, revealing a strong correlation between in situ membrane distribution and in vitro determined activity measurements. Here we describe the steps for tissue preparation and calpain activity measurement in the Drosophila embryo.
Sigma-Aldrich
E-64, protease inhibitor
Pricing and availability is not currently available.
Sigma-Aldrich
N-Succinyl-Leu-Tyr-7-amido-4-methylcoumarin, calpain substrate
Pricing and availability is not currently available.