Merck
  • Red/Near-Infrared Thermally Activated Delayed Fluorescence OLEDs with Near 100 % Internal Quantum Efficiency.
Red/Near-Infrared Thermally Activated Delayed Fluorescence OLEDs with Near 100 % Internal Quantum Efficiency.
Angewandte Chemie (International ed. in English) (2019-07-18)
Jia-Xiong Chen, Wen-Wen Tao, Wen-Cheng Chen, Ya-Fang Xiao, Kai Wang, Chen Cao, Jia Yu, Shengliang Li, Feng-Xia Geng, Chihaya Adachi, Chun-Sing Lee, Xiao-Hong Zhang
ABSTRACT
Developing red thermally activated delayed fluorescence (TADF) emitters, attainable for both high-efficient red organic light-emitting diodes (OLEDs) and non-doped deep red/near-infrared (NIR) OLEDs, is challenging. Now, two red emitters, BPPZ-PXZ and mDPBPZ-PXZ, with twisted donor-acceptor structures were designed and synthesized to study molecular design strategies of high-efficiency red TADF emitters. BPPZ-PXZ employs the strictest molecular restrictions to suppress energy loss and realizes red emission with a photoluminescence quantum yield (ΦPL ) of 100±0.8 % and external quantum efficiency (EQE) of 25.2 % in a doped OLED. Its non-doped OLED has an EQE of 2.5 % owing to unavoidable intermolecular π-π interactions. mDPBPZ-PXZ releases two pyridine substituents from its fused acceptor moiety. Although mDPBPZ-PXZ realizes a lower EQE of 21.7 % in the doped OLED, its non-doped device shows a superior EQE of 5.2 % with a deep red/NIR emission at peak of 680 nm.
Sigma-Aldrich
1,3,5-Tri[(3-pyridyl)-phen-3-yl]benzene, ≥99% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
4,4′-Bis(carbazol-9-yl)biphenyl, ≥99% (HPLC)
Pricing and availability is not currently available.