Merck
  • Central nervous system-infiltrated immune cells induce calcium increase in astrocytes via astroglial purinergic signaling.
Central nervous system-infiltrated immune cells induce calcium increase in astrocytes via astroglial purinergic signaling.
Journal of neuroscience research (2020-08-18)
Dunja D Bijelić, Katarina D Milićević, Milica N Lazarević, Djordje M Miljković, Jelena J Bogdanović Pristov, Danijela Z Savić, Branka B Petković, Pavle R Andjus, Miljana B Momčilović, Ljiljana M Nikolić
ABSTRACT
Interaction between autoreactive immune cells and astroglia is an important part of the pathologic processes that fuel neurodegeneration in multiple sclerosis. In this inflammatory disease, immune cells enter into the central nervous system (CNS) and they spread through CNS parenchyma, but the impact of these autoreactive immune cells on the activity pattern of astrocytes has not been defined. By exploiting naïve astrocytes in culture and CNS-infiltrated immune cells (CNS IICs) isolated from rat with experimental autoimmune encephalomyelitis (EAE), here we demonstrate previously unrecognized properties of immune cell-astrocyte interaction. We show that CNS IICs but not the peripheral immune cell application, evokes a rapid and vigorous intracellular Ca2+ increase in astrocytes by promoting glial release of ATP. ATP propagated Ca2+ elevation through glial purinergic P2X7 receptor activation by the hemichannel-dependent nucleotide release mechanism. Astrocyte Ca2+ increase is specifically triggered by the autoreactive CD4+ T-cell application and these two cell types exhibit close spatial interaction in EAE. Therefore, Ca2+ signals may mediate a rapid astroglial response to the autoreactive immune cells in their local environment. This property of immune cell-astrocyte interaction may be important to consider in studies interrogating CNS autoimmune disease.
Sigma-Aldrich
Monoclonal Anti-CD4 antibody produced in mouse, clone OX-35, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.