Merck
  • Sex-dependent pharmacological profiles of the synthetic cannabinoid MMB-Fubinaca.
Sex-dependent pharmacological profiles of the synthetic cannabinoid MMB-Fubinaca.
Addiction biology (2020-08-04)
José F Oliveira da Cruz, Christina Ioannidou, Antonio C Pagano Zottola, Carolina Muguruza, Paula Gomez-Sotres, Monica Fernandez, Luis F Callado, Giovanni Marsicano, Arnau Busquets-Garcia
ABSTRACT
Synthetic cannabinoids have emerged as novel psychoactive substances with damaging consequences for public health. They exhibit high affinity at the cannabinoid type-1 (CB1 ) receptor and produce similar and often more potent effects as other CB1 receptor agonists. However, we are still far from a complete pharmacological understanding of these compounds. In this study, by using behavioral, molecular, pharmacological, and electrophysiological approaches, we aimed at characterizing several in vitro and in vivo pharmacological effects of the synthetic cannabinoid MMB-Fubinaca (also known as AMB-Fubinaca or FUB-AMB), a particular synthetic cannabinoid. MMB-Fubinaca stimulates CB1 receptor-mediated functional coupling to G-proteins in mouse and human brain preparations in a similar manner as the CB1 receptor agonist WIN55,512-2 but with a much greater potency. Both drugs similarly activate the CB1 receptor-dependent extracellular signal-regulated kinase (ERK) pathway. Notably, in vivo administration of MMB-Fubinaca in mice induced greater behavioral and electrophysiological effects in male than in female mice in a CB1 receptor-dependent manner. Overall, these data provide a solid pharmacological profiling of the effects of MMB-Fubinaca and important information about the mechanisms of action underlying its harmful impact in humans. At the same time, they reinforce the significant sexual dimorphism of cannabinoid actions, which will have to be taken into account in future animal and clinical studies.
Sigma-Aldrich
(R)-(+)-WIN 55,212-2 mesylate salt, ≥98% (HPLC)