Merck
  • A Cell-Free Assay for Rapid Screening of Inhibitors of hACE2-Receptor-SARS-CoV-2-Spike Binding.
A Cell-Free Assay for Rapid Screening of Inhibitors of hACE2-Receptor-SARS-CoV-2-Spike Binding.
ACS synthetic biology (2022-04-05)
Nanami Kikuchi, Or Willinger, Naor Granik, Reut Gal, Noa Navon, Shanny Ackerman, Ella Samuel, Tomer Antman, Noa Katz, Sarah Goldberg, Roee Amit
ABSTRACT
We present a cell-free assay for rapid screening of candidate inhibitors of protein binding, focusing on inhibition of the interaction between the SARS-CoV-2 Spike receptor binding domain (RBD) and human angiotensin-converting enzyme 2 (hACE2). The assay has two components: fluorescent polystyrene particles covalently coated with RBD, termed virion-particles (v-particles), and fluorescently labeled hACE2 (hACE2F) that binds the v-particles. When incubated with an inhibitor, v-particle-hACE2F binding is diminished, resulting in a reduction in the fluorescent signal of bound hACE2F relative to the noninhibitor control, which can be measured via flow cytometry or fluorescence microscopy. We determine the amount of RBD needed for v-particle preparation, v-particle incubation time with hACE2F, hACE2F detection limit, and specificity of v-particle binding to hACE2F. We measure the dose response of the v-particles to known inhibitors. Finally, utilizing an RNA-binding protein tdPP7 incorporated into hACE2F, we demonstrate that RNA-hACE2F granules trap v-particles effectively, providing a basis for potential RNA-hACE2F therapeutics.
Sigma-Aldrich
GST protein, tag-free, recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution