Merck
  • Discovery of a Novel BCL-XL PROTAC Degrader with Enhanced BCL-2 Inhibition.
Discovery of a Novel BCL-XL PROTAC Degrader with Enhanced BCL-2 Inhibition.
Journal of medicinal chemistry (2021-09-18)
Pratik Pal, Dinesh Thummuri, Dongwen Lv, Xingui Liu, Peiyi Zhang, Wanyi Hu, Saikat K Poddar, Nan Hua, Sajid Khan, Yaxia Yuan, Xuan Zhang, Daohong Zhou, Guangrong Zheng
ABSTRACT
BCL-XL and BCL-2 are important targets for cancer treatment. BCL-XL specific proteolysis-targeting chimeras (PROTACs) have been developed to circumvent the on-target platelet toxicity associated with BCL-XL inhibition. However, they have minimal effects on cancer cells that are dependent on BCL-2 or both BCL-XL and BCL-2. Here we report a new series of BCL-PROTACs. The lead PZ703b exhibits high potency in inducing BCL-XL degradation and in inhibiting but not degrading BCL-2, showing a hybrid dual-targeting mechanism of action that is unprecedented in a PROTAC molecule. As a result, PZ703b is highly potent in killing BCL-XL dependent, BCL-2 dependent, and BCL-XL/BCL-2 dual-dependent cells in an E3 ligase (VHL)-dependent fashion. We further found that PZ703b forms stable {BCL-2:PROTAC:VCB} ternary complexes in live cells that likely contribute to the enhanced BCL-2 inhibition by PZ703b. With further optimization, analogues of PZ703b could potentially be developed as effective antitumor agents by co-targeting BCL-XL and BCL-2.
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Sigma-Aldrich
Bcl-xL Active human, recombinant, expressed in E. coli, ≥90% (SDS-PAGE)
Sigma-Aldrich
Bcl-2 Active human, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)