Merck
  • Production of Multi-Subtype Influenza Virus-Like Particles by Molecular Fusion with BAFF or APRIL for Vaccine Development.
Production of Multi-Subtype Influenza Virus-Like Particles by Molecular Fusion with BAFF or APRIL for Vaccine Development.
Methods in molecular biology (Clifton, N.J.) (2020-11-14)
Ting-Hsuan Chen, Jo-Yu Hong, Chia-Chyi Liu, Chung-Chu Chen, Jia-Tsrong Jan, Suh-Chin Wu
ABSTRACT
Virus-like particle (VLP) technology is an alternative platform for developing vaccines to combat seasonal and pandemic influenza. Influenza VLPs are non-infectious nanoparticles that can elicit effective vaccine immunogenicity in hosts. B-cell-activating factor (BAFF, or BLyS) and a proliferation-inducing ligand (APRIL) are members of the tumor necrosis factor (TNF) superfamily of cytokines. Both BAFF and APRIL are homotrimers that interact with homotrimeric receptors. Here, we report a method of the production of influenza VLPs by molecular incorporation with BAFF or APRIL homotrimers to interact with their receptors. We engineered the VLPs by direct fusion of BAFF or APRIL to the transmembrane anchored domain of the hemagglutinin (HA) gene. We also describe procedures for the production of BAFF-VLPs containing H5H7 and H1H5H7 for multi-subtype vaccine development.
Sigma-Aldrich
Fetuin from fetal bovine serum, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Lectin from Arachis hypogaea (peanut), peroxidase conjugate, lyophilized powder
Pricing and availability is not currently available.