Merck
  • Immunohistochemistry and In Situ Hybridization in the Developing Chicken Brain.
Immunohistochemistry and In Situ Hybridization in the Developing Chicken Brain.
Methods in molecular biology (Clifton, N.J.) (2019-09-26)
Richard P Tucker, Tatsuto Ishimaru, Qizhi Gong
ABSTRACT
One of the first steps in studies of gene function is the spatiotemporal analysis of patterns of gene expression. Indirect immunohistochemistry is a method that allows the detection of a protein of interest by incubating a histological section with an antibody or antiserum raised against the protein, and then localizing this primary antibody with a tagged secondary antibody. To determine the cellular source of a protein of interest, or if a specific antibody is not available, specific transcripts can be localized using in situ hybridization. A histological section is incubated with a labeled RNA probe that is complementary to the target transcript; after hybridization with the target transcript the labeled RNA probe can be identified with an antibody. Here we describe materials and methods used to perform basic indirect immunohistochemistry and in situ hybridization on frozen sections through the developing chicken brain, emphasizing controls and potential problems that may be encountered.
Sigma-Aldrich
Proteinase K from Tritirachium album
Pricing and availability is not currently available.