Merck
  • Exosomes from mesenchymal stem cells overexpressing MIF enhance myocardial repair.
Exosomes from mesenchymal stem cells overexpressing MIF enhance myocardial repair.
Journal of cellular physiology (2020-01-22)
Xiaolin Liu, Xin Li, Wenwu Zhu, Yuelin Zhang, Yimei Hong, Xiaoting Liang, Baohan Fan, Hongyan Zhao, Haiwei He, Fengxiang Zhang
ABSTRACT
Accumulating evidence has shown that mesenchymal stem cell (MSC)-derived exosomes (exo) mediate cardiac repair following myocardial infarction (MI). Macrophage migration inhibitory factor (MIF), a proinflammatory cytokine, plays a critical role in regulating cell homeostasis. This study aimed to investigate the cardioprotective effects of exo secreted from bone marrow-MSCs (BM-MSCs) overexpressing MIF in a rat model of MI. MIF plasmid was transducted in BM-MSCs. Exo were isolated from the supernatants of BM-MSCs and MIF-BM-MSCs, respectively. The morphology of mitochondria in neonatal mice cardiomyocytes (NRCMs) was determined by MitoTracker staining. The apoptosis of NRCMs was examined by deoxynucleotidyl transferase-mediated dUTP nick end-labeling. BM-MSC-exo and MIF-BM-MSC-exo were intramuscularly injected into the peri-infarct region in a rat model of MI. The heart function of rats was assessed by echocardiography. The expression of MIF was greatly enhanced in MIF-BM-MSCs compared with BM-MSCs. Both BM-MSC-exo and MIF-BM-MSC-exo expressed CD63 and CD81. NRCMs treated with MIF-BM-MSC-exo exhibited less mitochondrial fragmentation and cell apoptosis under hypoxia/serum deprivation (H/SD) challenge than those treated with BM-MSC-exo via activating adenosine 5'-monophosphate-activated protein kinase signaling. Moreover, these effects were partially abrogated by Compound C. Injection of BM-MSC-exo or MIF-BM-MSC-exo greatly restored heart function in a rat model of MI. Compared with BM-MSC-exo, injection of MIF-BM-MSC-exo was associated with enhanced heart function, reduced heart remodeling, less cardiomyocyte mitochondrial fragmentation, reactive oxygen species generation, and apoptosis. Our study reveals a new mechanism of MIF-BM-MSC-exo-based therapy for MI and provides a novel strategy for cardiovascular disease treatment.
Sigma-Aldrich
Anti-FIS1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.