Merck
  • The effects of mesenchymal stem cell mitochondrial transplantation on doxorubicin-mediated nephrotoxicity in rats.
The effects of mesenchymal stem cell mitochondrial transplantation on doxorubicin-mediated nephrotoxicity in rats.
Journal of biochemical and molecular toxicology (2020-09-02)
Gokhan Burcin Kubat, Mehmet Ozler, Oner Ulger, Ozgur Ekinci, Ozbeyen Atalay, Ertugrul Celik, Mukerrem Safali, Murat Timur Budak
ABSTRACT
The effect of dysfunctional mitochondria in several cell pathologies has been reported in renal diseases, including diabetic nephropathy and acute kidney injury. Previous studies have reported that mitochondrial transplantation provided surprising results in myocardial and liver ischemia, as well as in Parkinson's disease. We aimed to investigate the beneficial effects of isolated mitochondria transplantation from mesenchymal stem cells (MSCs) in vivo, to mitigate renal damage that arises from doxorubicin-mediated nephrotoxicity and its action mechanism. In this study, a kidney model of doxorubicin-mediated nephrotoxicity was used and isolated mitochondria from MSCs were transferred to the renal cortex of rats. The findings showed that the rate of isolated mitochondria from MSCs maintains sufficient membrane integrity, and was associated with a beneficial renal therapeutic effect. Following doxorubicin-mediated renal injury, isolated mitochondria or vehicle infused into the renal cortex and rats were monitored for five days. This study found that mitochondrial transplantation decreased cellular oxidative stress and promoted regeneration of tubular cells after renal injury (P < .001, P = .009). Moreover, mitochondrial transplantation reduced protein accumulation of tubular cells and reversed renal deficits (P = .01, P < .001). Mitochondrial transplantation increased Bcl-2 levels, and caspase-3 levels decreased in injured renal cells (P < .015, P < .001). Our results provide a direct link between mitochondria dysfunction and doxorubicin-mediated nephrotoxicity and suggest a therapeutic effect of transferring isolated mitochondria obtained from MSCs against renal injury. To our knowledge, this study is the first study in the literature that showed good therapeutic effects of mitochondrial transplantation in a nephrotoxicity model, which is under-researched.
Sigma-Aldrich
Rat Mesenchymal Stem Cells (Bone Marrow), These primary bone marrow derived Rat Mesenchymal Stem Cells isolated from the iliac crest of adult Fisher 344 rats provide a convenient source of multipotent rat mesenchymal stem cells.