- Levels of matrix metalloproteinase-2 and metalloproteinase-9 in the cerebrospinal fluid of dogs with visceral leishmaniasis.
Levels of matrix metalloproteinase-2 and metalloproteinase-9 in the cerebrospinal fluid of dogs with visceral leishmaniasis.
Inflammation causes increases in the level of matrix metalloproteinases (MMPs), which, in central nervous system (CNS), are associated with neuroinflammation and disruption of blood-brain barrier. Analysis of cerebrospinal fluid (CSF) is pivotal for detecting diseases in CNS and, although a specific diagnosis may not be achieved, this analysis is helpful to confirm the diagnosis or to rule out relevant differential diagnoses. This study examined the levels of MMP-2 and MMP-9 in the CSF of dogs using gelatin zymography to verify possible alterations in these enzymes during natural systemic infection with Leishmania chagasi. Latent and active forms of MMP-2 were detected in some dogs of both groups, with high levels in the control group. In contrast, latent and active forms of MMP-9 were detected only in some animals with leishmaniasis. These results clearly demonstrate that MMP-9 is elevated in CSF of dogs with visceral leishmaniasis (VL). Although these results are preliminary, they suggest that MMP-9 might play a role in disruption of blood-brain barrier and/or blood-CSF barrier. While the presence of MMPs in CSF is not a condition exclusive to VL, their presence and persistence in CSF supports the hypothesis of an inflammatory state within CNS of dogs with VL.