Merck
U2.3 Precursor Small Nuclear RNA in vitro Processing Assay.
Bio-protocol (2021-10-05)
Chan Lin, Yujie Feng, Xueyan Peng, Jiaming Wu, Weili Wang, Yunfeng Liu
ABSTRACT
Small nuclear RNAs (snRNAs) are vital for eukaryotic cell activities and play important roles in pre-mRNA splicing. The molecular mechanism underlying the transcription of snRNA, regulated via upstream/downstream cis-elements and relevant trans-elements, has been investigated in detail using cell-free extracts. However, the processing of precursor snRNA (pre-snRNA), which is required by 3' end maturation of pre-snRNA, remains unclear as a proper processing assay is difficult to develop in vitro. Here, we present an in vitro method using synthetic labeled RNA as substrates to study the 3' cleavage of pre-snRNA.
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Phenylmethylsulfonyl Fluoride, Phenylmethylsulfonyl Fluoride, CAS 329-98-6, is an irreversible inhibitor of serine proteases. It causes sulfonylation of the active-site serine residues.