Merck
Multimodal small-molecule screening for human prion protein binders.
The Journal of biological chemistry (2020-07-30)
Andrew G Reidenbach, Michael F Mesleh, Dominick Casalena, Sonia M Vallabh, Jayme L Dahlin, Alison J Leed, Alix I Chan, Dmitry L Usanov, Jenna B Yehl, Christopher T Lemke, Arthur J Campbell, Rishi N Shah, Om K Shrestha, Joshua R Sacher, Victor L Rangel, Jamie A Moroco, Murugappan Sathappa, Maria Cristina Nonato, Kong T Nguyen, S Kirk Wright, David R Liu, Florence F Wagner, Virendar K Kaushik, Douglas S Auld, Stuart L Schreiber, Eric Vallabh Minikel
ABSTRACT
Prion disease is a rapidly progressive neurodegenerative disorder caused by misfolding and aggregation of the prion protein (PrP), and there are currently no therapeutic options. PrP ligands could theoretically antagonize prion formation by protecting the native protein from misfolding or by targeting it for degradation, but no validated small-molecule binders have been discovered to date. We deployed a variety of screening methods in an effort to discover binders of PrP, including 19F-observed and saturation transfer difference (STD) NMR spectroscopy, differential scanning fluorimetry (DSF), DNA-encoded library selection, and in silico screening. A single benzimidazole compound was confirmed in concentration-response, but affinity was very weak (K d > 1 mm), and it could not be advanced further. The exceptionally low hit rate observed here suggests that PrP is a difficult target for small-molecule binders. Whereas orthogonal binder discovery methods could yield high-affinity compounds, non-small-molecule modalities may offer independent paths forward against prion disease.
Sigma-Aldrich
Overnight Express Autoinduction System 1 - Novagen, The Overnight Express Autoinduction System allows the induction of protein expression without monitoring cell density and without conventional induction with IPTG.
Pricing and availability is not currently available.