Merck
  • Insulin activates intracellular transport of lipid droplets to release triglycerides from the liver.
Insulin activates intracellular transport of lipid droplets to release triglycerides from the liver.
The Journal of cell biology (2019-10-13)
Mukesh Kumar, Srikant Ojha, Priyanka Rai, Alaumy Joshi, Siddhesh S Kamat, Roop Mallik
ABSTRACT
Triglyceride-rich lipid droplets (LDs) are catabolized with high efficiency in hepatocytes to supply fatty acids for producing lipoprotein particles. Fasting causes a massive influx of adipose-derived fatty acids into the liver. The liver in the fasted state is therefore bloated with LDs but, remarkably, still continues to secrete triglycerides at a constant rate. Here we show that insulin signaling elevates phosphatidic acid (PA) dramatically on LDs in the fed state. PA then signals to recruit kinesin-1 motors, which transport LDs to the peripherally located smooth ER inside hepatocytes, where LDs are catabolized to produce lipoproteins. This pathway is down-regulated homeostatically when fasting causes insulin levels to drop, thus preventing dangerous elevation of triglycerides in the blood. Further, we show that a specific peptide against kinesin-1 blocks triglyceride secretion without any apparent deleterious effects on cells. Our work therefore reveals fundamental mechanisms that maintain lipid homeostasis across metabolic states and leverages this knowledge to propose a molecular target against hyperlipidemia.
Sigma-Aldrich
VU0155056, ≥98% (HPLC)