Merck
  • NFIA determines the cis-effect of genetic variation on Ucp1 expression in murine thermogenic adipocytes.
NFIA determines the cis-effect of genetic variation on Ucp1 expression in murine thermogenic adipocytes.
iScience (2022-07-26)
Yuta Hiraike, Shuichi Tsutsumi, Takahito Wada, Misato Oguchi, Kaede Saito, Masahiro Nakamura, Satoshi Ota, Michinori Koebis, Harumi Nakao, Atsu Aiba, Gaku Nagano, Haruya Ohno, Kenji Oki, Masayasu Yoneda, Takashi Kadowaki, Hiroyuki Aburatani, Hironori Waki, Toshimasa Yamauchi
ABSTRACT
Thermogenic brown and beige adipocytes counteract obesity by enhancing energy dissipation via uncoupling protein-1 (Ucp1). However, the effect of genetic variation on these cells, a major source of disease susceptibility, has been less well studied. Here we examined beige adipocytes from obesity-prone C57BL/6J (B6) and obesity-resistant 129X1/SvJ (129) mouse strains and identified a cis-regulatory variant rs47238345 that is responsible for differential Ucp1 expression. The alternative T allele of rs47238345 at the Ucp1 -12kb enhancer in 129 facilitates the allele-specific binding of nuclear factor I-A (NFIA) to mediate allele-specific enhancer-promoter interaction and Ucp1 transcription. Furthermore, CRISPR-Cas9/Cpf1-mediated single nucleotide polymorphism (SNP) editing of rs47238345 resulted in increased Ucp1 expression. We also identified Lim homeobox protein 8 (Lhx8), whose expression is higher in 129 than in B6, as a trans-acting regulator of Ucp1 in mice and humans. These results demonstrate the cis- and trans-acting effects of genetic variation on Ucp1 expression that underlie phenotypic diversity.
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NFIA antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)