Merck
  • Synaptamide Modulates Astroglial Activity in Mild Traumatic Brain Injury.
Synaptamide Modulates Astroglial Activity in Mild Traumatic Brain Injury.
Marine drugs (2022-08-26)
Arina Ponomarenko, Anna Tyrtyshnaia, Darya Ivashkevich, Ekaterina Ermolenko, Inessa Dyuizen, Igor Manzhulo
ABSTRACT
At present, the study of the neurotropic activity of polyunsaturated fatty acid ethanolamides (N-acylethanolamines) is becoming increasingly important. N-docosahexaenoylethanolamine (synaptamide, DHEA) is a highly active metabolite of docosahexaenoic acid (DHA) with neuroprotective, synaptogenic, neuritogenic, and anti-inflammatory properties in the nervous system. Synaptamide tested in the present study was obtained using a chemical modification of DHA isolated from squid Berryteuthis magister liver. The results of this study demonstrate the effects of synaptamide on the astroglial response to injury in the acute (1 day) and chronic (7 days) phases of mild traumatic brain injury (mTBI) development. HPLC-MS study revealed several times increase of synaptamide concentration in the cerebral cortex and serum of experimental animals after subcutaneous administration (10 mg/kg/day). Using immunohistochemistry, it was shown that synaptamide regulates the activation of GFAP- and S100β-positive astroglia, reduce nNOS-positive immunostaining, and stimulates the secretion of neurotrophin BDNF. Dynamics of superoxide dismutase production in synaptamide treatment confirm the antioxidant efficacy of the test compound. We found a decrease in TBI biomarkers such as GFAP, S100β, and IL-6 in the blood serum of synaptamide-treated experimental animals using Western blot analysis. The results indicate the high therapeutic potential of synaptamide in reducing the severity of the brain damage consequences.
Sigma-Aldrich
SOD Assay Kit, sufficient for 500 tests
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-nNOS/NOS I Antibody, 0.5 mg/mL, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Superoxide Dismutase bovine, recombinant, expressed in E. coli, lyophilized powder, ≥2500 units/mg protein, ≥90% (SDS-PAGE)
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-GFAP antibody produced in mouse, Prestige Antibodies® Powered by Atlas Antibodies, clone CL2713, purified immunoglobulin, buffered aqueous glycerol solution
Pricing and availability is not currently available.