Merck
  • Glucose-induced cAMP elevation in β-cells involves amplification of constitutive and glucagon-activated GLP-1 receptor signalling.
Glucose-induced cAMP elevation in β-cells involves amplification of constitutive and glucagon-activated GLP-1 receptor signalling.
Acta physiologica (Oxford, England) (2020-12-29)
Hongyan Shuai, Yunjian Xu, Parvin Ahooghalandari, Anders Tengholm
ABSTRACT
cAMP typically signals downstream of Gs -coupled receptors and regulates numerous cell functions. In β-cells, cAMP amplifies Ca2+ -triggered exocytosis of insulin granules. Glucose-induced insulin secretion is associated with Ca2+ - and metabolism-dependent increases of the sub-plasma-membrane cAMP concentration ([cAMP]pm ) in β-cells, but potential links to canonical receptor signalling are unclear. The aim of this study was to clarify the role of glucagon-like peptide-1 receptors (GLP1Rs) for glucose-induced cAMP signalling in β-cells. Total internal reflection microscopy and fluorescent reporters were used to monitor changes in cAMP, Ca2+ and ATP concentrations as well as insulin secretion in MIN6 cells and mouse and human β-cells. Insulin release from mouse and human islets was also measured with ELISA. The GLP1R antagonist exendin-(9-39) (ex-9) prevented both GLP1- and glucagon-induced elevations of [cAMP]pm , consistent with GLP1Rs being involved in the action of glucagon. This conclusion was supported by lack of unspecific effects of the antagonist in a reporter cell-line. Ex-9 also suppressed IBMX- and glucose-induced [cAMP]pm elevations. Depolarization with K+ triggered Ca2+ -dependent [cAMP]pm elevation, an effect that was amplified by high glucose. Ex-9 inhibited both the Ca2+ and glucose-metabolism-dependent actions on [cAMP]pm . The drug remained effective after minimizing paracrine signalling by dispersing the islets and it reduced basal [cAMP]pm in a cell-line heterologously expressing GLP1Rs, indicating that there is constitutive GLP1R signalling. The ex-9-induced reduction of [cAMP]pm in glucose-stimulated β-cells was paralleled by suppression of insulin secretion. Agonist-independent and glucagon-stimulated GLP1R signalling in β-cells contributes to basal and glucose-induced cAMP production and insulin secretion.
Sigma-Aldrich
Ready-to-Assay Glucagon Receptor Frozen Cells, Human glucagon / GCG GPCR frozen cells for Calcium flux Assays, ligand binding FLIPR Assays.