Merck
  • POLYRETINA restores light responses in vivo in blind Göttingen minipigs.
POLYRETINA restores light responses in vivo in blind Göttingen minipigs.
Nature communications (2022-06-28)
Paola Vagni, Marta Jole Ildelfonsa Airaghi Leccardi, Charles-Henri Vila, Elodie Geneviève Zollinger, Golnaz Sherafatipour, Thomas J Wolfensberger, Diego Ghezzi
ABSTRACT
Retinal prostheses hold the potential for artificial vision in blind people affected by incurable diseases of the outer retinal layer. Available technologies provide only a small field of view: a significant limitation for totally blind people. To overcome this problem, we recently proposed a large and high-density photovoltaic epiretinal device, known as POLYRETINA. Here, we report the in vivo assessment of POLYRETINA. First, we characterise a model of chemically-induced blindness in Göttingen minipigs. Then, we develop and test a minimally invasive injection procedure to insert the large epiretinal implant into the eye. Last, we show that POLYRETINA restores light-evoked cortical responses in blind animals at safe irradiance levels. These results indicate that POLYRETINA holds the potential for artificial vision in totally blind patients affected by retinitis pigmentosa.
Sigma-Aldrich
Anti-Rabbit IgG (H+L), CF 488A antibody produced in goat, ~2 mg/mL, affinity isolated antibody
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Opsin Antibody, blue, Chemicon®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Monoclonal Anti-Na+/K+ ATPase (α3 Subunit) antibody produced in mouse, 1 mg/mL, clone XVIF9-G10, ascites fluid
Pricing and availability is not currently available.