Merck
  • Asparaginase induces selective dose- and time-dependent cytotoxicity, apoptosis, and reduction of NFκB expression in oral cancer cells.
Asparaginase induces selective dose- and time-dependent cytotoxicity, apoptosis, and reduction of NFκB expression in oral cancer cells.
Clinical and experimental pharmacology & physiology (2020-01-17)
Gabriel Álvares Borges, Silvia Taveira Elias, Tassiana Souza De Araujo, Paula Monteiro Souza, Carlos Henrique Viesi Nascimento-Filho, Rogerio M Castilho, Cristiane H Squarize, Pérola de Oliveira Magalhães, Eliete Neves Silva Guerra
ABSTRACT
Asparaginase is fundamental to the treatment of haematological malignancies. However, little has been studied on the effects that asparaginase could exert on solid tumours. Thus, this study aimed to evaluate the effects of asparaginase on an oral carcinoma cell line. The cytotoxicity of asparaginase in SCC-9 (tongue squamous cell carcinoma) and HaCaT (human keratinocyte) cell lines was evaluated with MTT cell viability assay. The cells were treated with asparaginase at 0.04, 0.16, 0.63, 1.0, 1.5, 2.5, and 5.0 IU/mL. Dose-response curves and IC50 values were obtained and the Tumour Selectivity Index (TSI) was calculated. The effect of asparaginase on procaspase-3 and nuclear factor κB (NFκB) expression was evaluated with western blot because it was reported that the overexpression of NFκB has been shown to contribute to tumour cell survival, proliferation, and migration. Caspase 3/7 staining was performed to identify cell death using flow cytometry. Effective asparaginase concentrations were lower for SCC-9 cells when compared to HaCaT cells. The cytotoxicity results at 48 and 72 hours were significantly different for SCC-9 cells. The TSI indicated that asparaginase was selective for the tumour cells. A decrease in procaspase-3 and NFκB protein levels was observed in SCC-9 cells. Furthermore, asparaginase resulted in significant apoptosis after 48 and 72 hours. Based on these results, asparaginase was cytotoxic in a dose- and time-dependent manner, induces apoptosis, and reduces NFκB expression in oral cancer cells. These results encourage further studies on the effectiveness of this enzyme as a treatment for solid tumours, especially head and neck cancer.
Sigma-Aldrich
Asparaginase from Escherichia coli, lyophilized powder, 100-300 units/mg protein (biuret)
Pricing and availability is not currently available.