Merck
  • Impaired T cell proliferation and zeta chain phosphorylation after stimulation with staphylococcal enterotoxin-B in hemodialysis patients.
Impaired T cell proliferation and zeta chain phosphorylation after stimulation with staphylococcal enterotoxin-B in hemodialysis patients.
Nephron. Clinical practice (2004-01-31)
Theodoros Eleftheriadis, Konstantinos Papazisis, Alexandros Kortsaris, Georgios Vayonas, Sultana Voyatzi, Vasilios Vargemezis
ABSTRACT
Patients on regular hemodialysis treatment are in an immunodeficiency state. Several studies have shown defective T cell proliferation after stimulation with various agents. Staphylococcal enterotoxin B (SEB) is a MHC-dependent superantigen that triggers proliferation of a large proportion of T cells. T cell activation after stimulation with SEB parallels normal T cell signal transduction. An important and early event in this transduction pathway is the phosphorylation of the zeta chain. In this study, T cell proliferation and zeta chain phosphorylation after stimulation with SEB were evaluated. Peripheral blood mononuclear cells (PBMCs) from 24 patients and 14 healthy individuals were isolated and cultured with or without stimulation with SEB (1 ng/ml). Cell proliferation was estimated by immunoenzymatic measurement of bromodeoxyuridine uptake. PBMCs from 8 patients and 6 healthy individuals were isolated and pulsed for 2 min with or without SEB (10 microg/ml). Zeta chain phosphorylation was estimated by immunoprecipitation and immunoblotting with antiphosphotyrosine antibody. Lymphocyte proliferation index after SEB stimulation was lower in hemodialyzed patients. Stimulation of T cells with SEB also resulted in a lower zeta chain phosphorylation in hemodialyzed patients. Lymphocyte proliferation after MHC-dependent stimulation is impaired in hemodialyzed patients. This proliferation defect is due to impaired zeta chain phosphorylation.
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution