- Therapeutic use of α2-antiplasmin as an antifibrinolytic and hemostatic agent in surgery and regenerative medicine.
Therapeutic use of α2-antiplasmin as an antifibrinolytic and hemostatic agent in surgery and regenerative medicine.
The biomaterial fibrin is widely used as a clinical tissue sealant in surgery. In preclinical research, fibrin is also extensively studied as a carrier material for growth factor delivery. In these applications, premature fibrin degradation leads to recurrent bleeding, tissue dehiscence and limited regenerative efficacy. Therefore, fibrinolysis inhibitors have been added to clinical fibrin formulations, for example the bovine-derived serine protease inhibitor aprotinin. Aprotinin is additionally used as a hemostatic agent to prevent excessive bleeding during surgery, in this case protecting endogenous fibrin clots. Nevertheless, aprotinin use has been associated with serious safety issues. Here, we explore the use the human physiological fibrinolysis inhibitor α2-antiplasmin (α2PI) as a substitute for aprotinin. We evaluate the efficacy of α2PI in the three main applications of aprotinin. We first showed that recombinant α2PI can successfully prolong the durability of fibrin biomaterials as compared to aprotinin in a model of subcutaneous implantation in mice mimicking application as a tissue sealant. We then used α2PI to enhance the delivery of engineered vascular endothelial growth factor (VEGF)-A and platelet-derived growth factor (PDGF)-BB in fibrin in promoting diabetic wound healing, which lead to improved wound closure, granulation tissue formation and angiogenesis. Lastly, we demonstrated that α2PI can be as effective as aprotinin as an intravenous hemostatic agent to prevent blood loss, using a tail-vein bleeding model in mice. Therefore, we believe that engineering fibrin biomaterials or endogenous fibrin with α2PI can have a strong impact in surgery and regenerative medicine by providing a competitive substitute to aprotinin that is of human origin.