Merck
  • A Catalytically Independent Function of Human DNA Polymerase Kappa Controls the Stability and Abundance of Checkpoint Kinase 1.
A Catalytically Independent Function of Human DNA Polymerase Kappa Controls the Stability and Abundance of Checkpoint Kinase 1.
Molecular and cellular biology (2021-08-17)
Marina Dall'Osto, Laura Pierini, Nicolas Valery, Jean-Sébastien Hoffmann, Marie-Jeanne Pillaire
ABSTRACT
DNA polymerase kappa (Pol κ) has been well documented thus far for its specialized DNA synthesis activity during translesion replication, progression of replication forks through regions difficult to replicate, restart of stalled forks, and replication checkpoint efficiency. Pol κ is also required for the stabilization of stalled forks, although the mechanisms are poorly understood. In this study, we unveiled an unexpected role for Pol κ in controlling the stability and abundance of checkpoint kinase 1 (Chk1), an important actor for the replication checkpoint and fork stabilization. We found that loss of Pol κ decreased the Chk1 protein level in the nuclei of four human cell lines. Pol κ and not the other Y family polymerase members is required to maintain the Chk1 protein pool all along the cell cycle. We showed that Pol κ depletion affected the protein stability of Chk1 and protected it from proteasome degradation. Importantly, we also observed that the fork restart defects observed in Pol κ-depleted cells could be overcome by the reexpression of Chk1. Strikingly, this new function of Pol κ does not require its catalytic activity. We propose that Pol κ could contribute to the protection of stalled forks through Chk1 stability.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Sigma-Aldrich
Anti-Fibrillarin antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-α-Actinin Antibody, clone AT6/172, clone AT6/172, Upstate®, from mouse