- Intrathecal administration of thrombin inhibitor ameliorates cerebral vasospasm. Use of a drug delivery system releasing hirudin.
Intrathecal administration of thrombin inhibitor ameliorates cerebral vasospasm. Use of a drug delivery system releasing hirudin.
Cerebrovascular diseases (Basel, Switzerland) (2000-11-09)
A Kudo, M Suzuki, Y Kubo, M Watanabe, K Yoshida, M Doi, K Kuroda, A Ogawa
PMID11070371
ABSTRACT
The role of thrombin as a spasmogen after subarachnoid hemorrhage was evaluated using the intrathecally administered thrombin inhibitor hirudin, released from a drug delivery system (DDS) based on collagen in a canine vasospasm model. The DDS was implanted into the cisterna magna with autologous blood in the hirudin-treated group. The reduction in the angiographical diameter of the basilar artery was only 19% in the hirudin-treated group on day 7, showing a significant difference between hirudin-treated and nontreated groups (p < 0.01). These results suggest that thrombin is an important cause of vasospasm. The collagen DDS has great potential for treatment in the cerebrospinal fluid milieu.
Sigma-Aldrich
Hirudin, recombinant, expressed in unspecified host, ≥7,000 ATU/mg protein (ATU = antithrombin units)