Merck
  • Molecularly cleavable bioinks facilitate high-performance digital light processing-based bioprinting of functional volumetric soft tissues.
Molecularly cleavable bioinks facilitate high-performance digital light processing-based bioprinting of functional volumetric soft tissues.
Nature communications (2022-06-11)
Mian Wang, Wanlu Li, Jin Hao, Arthur Gonzales, Zhibo Zhao, Regina Sanchez Flores, Xiao Kuang, Xuan Mu, Terry Ching, Guosheng Tang, Zeyu Luo, Carlos Ezio Garciamendez-Mijares, Jugal Kishore Sahoo, Michael F Wells, Gengle Niu, Prajwal Agrawal, Alfredo Quiñones-Hinojosa, Kevin Eggan, Yu Shrike Zhang
ABSTRACT
Digital light processing bioprinting favors biofabrication of tissues with improved structural complexity. However, soft-tissue fabrication with this method remains a challenge to balance the physical performances of the bioinks for high-fidelity bioprinting and suitable microenvironments for the encapsulated cells to thrive. Here, we propose a molecular cleavage approach, where hyaluronic acid methacrylate (HAMA) is mixed with gelatin methacryloyl to achieve high-performance bioprinting, followed by selectively enzymatic digestion of HAMA, resulting in tissue-matching mechanical properties without losing the structural complexity and fidelity. Our method allows cellular morphological and functional improvements across multiple bioprinted tissue types featuring a wide range of mechanical stiffness, from the muscles to the brain, the softest organ of the human body. This platform endows us to biofabricate mechanically precisely tunable constructs to meet the biological function requirements of target tissues, potentially paving the way for broad applications in tissue and tissue model engineering.
Sigma-Aldrich
Monoclonal Anti-S-100 (β-Subunit) antibody produced in mouse, clone SH-B1, ascites fluid
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Synaptophysin Antibody, clone 2B21, ZooMAb® Rabbit Monoclonal, recombinant, expressed in HEK 293 cells
Pricing and availability is not currently available.