Merck
  • CIC-39Na reverses the thrombocytopenia that characterizes tubular aggregate myopathy.
CIC-39Na reverses the thrombocytopenia that characterizes tubular aggregate myopathy.
Blood advances (2022-06-14)
Celia Cordero-Sanchez, Emanuela Pessolano, Beatrice Riva, Mauro Vismara, Silvia Maria Grazia Trivigno, Nausicaa Clemente, Silvio Aprile, Federico Alessandro Ruffinatti, Paola Portararo, Nicoletta Filigheddu, Ivan Zaggia, Irene P Bhela, Marta Serafini, Tracey Pirali, Mario P Colombo, Mauro Torti, Sabina Sangaletti, Alessandra Bertoni, Armando A Genazzani
ABSTRACT
Store-operated Ca2+-entry is a cellular mechanism that governs the replenishment of intracellular stores of Ca2+ upon depletion caused by the opening of intracellular Ca2+-channels. Gain-of-function mutations of the 2 key proteins of store-operated Ca2+-entry, STIM1 and ORAI1, are associated with several ultra-rare diseases clustered as tubular aggregate myopathies. Our group has previously demonstrated that a mouse model bearing the STIM1 p.I115F mutation recapitulates the main features of the STIM1 gain-of-function disorders: muscle weakness and thrombocytopenia. Similar findings have been found in other mice bearing different mutations on STIM1. At present, no valid treatment is available for these patients. In the present contribution, we report that CIC-39Na, a store-operated Ca2+-entry inhibitor, restores platelet number and counteracts the abnormal bleeding that characterizes these mice. Subtle differences in thrombopoiesis were observed in STIM1 p.I115F mice, but the main difference between wild-type and STIM1 p.I115F mice was in platelet clearance and in the levels of platelet cytosolic basal Ca2+. Both were restored on treatment of animals with CIC-39Na. This finding paves the way to a pharmacological treatment strategy for thrombocytopenia in tubular aggregate myopathy patients.
Sigma-Aldrich
Apyrase from potato, recombinant, expressed in Pichia pastoris, ATPase ≥1000 units/mg protein, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Actin Antibody, clone C4, ascites fluid, clone C4, Chemicon®
Pricing and availability is not currently available.