Merck
  • NHP2 downregulation counteracts hTR-mediated activation of the DNA damage response at ALT telomeres.
NHP2 downregulation counteracts hTR-mediated activation of the DNA damage response at ALT telomeres.
The EMBO journal (2021-02-18)
Maya Raghunandan, Dan Geelen, Eva Majerova, Anabelle Decottignies
ABSTRACT
About 10% of cancer cells employ the "alternative lengthening of telomeres" (ALT) pathway instead of re-activating the hTERT subunit of human telomerase. The hTR RNA subunit is also abnormally silenced in some ALT+ cells not expressing hTERT, suggesting a possible negative non-canonical impact of hTR on ALT. Indeed, we show that ectopically expressed hTR reduces phosphorylation of ssDNA-binding protein RPA (p-RPAS33 ) at ALT telomeres by promoting the hnRNPA1- and DNA-PK-dependent depletion of RPA. The resulting defective ATR checkpoint signaling at telomeres impairs recruitment of the homologous recombination protein, RAD51. This induces ALT telomere fragility, increases POLD3-dependent C-circle production, and promotes the recruitment of the DNA damage marker 53BP1. In ALT+ cells that naturally retain hTR expression, NHP2 H/ACA ribonucleoprotein levels are downregulated, likely in order to restrain DNA damage response (DDR) activation at telomeres through reduced 53BP1 recruitment. This unexpected role of NHP2 is independent from hTR's non-canonical function in modulating telomeric p-RPAS33 . Collectively, our study shines new light on the interference between telomerase- and ALT-dependent pathways and unravels a crucial role for hTR and NHP2 in DDR regulation at ALT telomeres.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Vinculin Antibody, clone V284, clone V284, Upstate®, from mouse
Roche
cOmplete, Mini Protease Inhibitor Cocktail, Tablets provided in a glass vial
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-Histone H2A.X Antibody, serum, Upstate®
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys9) Antibody, Upstate®, from rabbit
Roche
TeloTAGGG Telomere Length Assay, sufficient for ≤50 reactions, kit of 1 (15 components), suitable for cell culture
Sigma-Aldrich
Anti-RNA polymerase II Antibody, clone CTD4H8, clone CTD4H8, Upstate®, from mouse
Sigma-Aldrich
Anti-acetyl-Histone H3 Antibody, from rabbit