Merck
  • Establishing a public health analytical service based on chemical methods for detecting and quantifying Pacific ciguatoxin in fish samples.
Establishing a public health analytical service based on chemical methods for detecting and quantifying Pacific ciguatoxin in fish samples.
Toxicon : official journal of the International Society on Toxinology (2009-08-04)
Ian Stewart, Geoffrey K Eaglesham, Sue Poole, Glenn Graham, Carl Paulo, Wasantha Wickramasinghe, Ross Sadler, Glen R Shaw
ABSTRACT
A referee analysis method for the detection and quantification of Pacific ciguatoxins in fish flesh has recently been established by the public health analytical laboratory for the State of Queensland, Australia. Fifty-six fish samples were analysed, which included 10 fillets purchased as negative controls. P-CTX-1 was identified in 27 samples, and P-CTX-2 and P-CTX-3 were found in 26 of those samples. The range of P-CTX-1 concentrations was 0.04-11.4 microg/kg fish flesh; coefficient of variation from 90 replicate analyses was 7.4%. A liquid chromatography/tandem mass spectrometry (HPLC-MS/MS) method utilising a rapid methanol extraction and clean-up is reliable and reproducible, with the detection limit at 0.03 microg/kg fish flesh. Some matrix effects are evident, with fish oil content a likely signal suppression factor. Species identification of samples by DNA sequence analysis revealed some evidence of fish substitution or inadvertent misidentification, which may have implications for the management and prevention of ciguatera poisoning. Blinded inspection of case notes from suspect ciguatera poisoning cases showed that reporting of ciguatera-related paraesthesias was highly predictable for the presence of ciguatoxins in analysed fish, with 13 of 14 expected cases having consumed fish that contained P-CTX-1 (p<0.001, Fishers Exact Test).
Product Description
Millipore
MultiScreen-PCRµ96 Filter Plate, Purification of PCR products
Pricing and availability is not currently available.