Merck
  • Establishment of an antibody specific for AMIGO2 improves immunohistochemical evaluation of liver metastases and clinical outcomes in patients with colorectal cancer.
Establishment of an antibody specific for AMIGO2 improves immunohistochemical evaluation of liver metastases and clinical outcomes in patients with colorectal cancer.
Diagnostic pathology (2022-02-01)
Keisuke Goto, Mitsuhiko Osaki, Runa Izutsu, Hiroshi Tanaka, Ryo Sasaki, Akimitsu Tanio, Hiroyuki Satofuka, Yasuhiro Kazuki, Manabu Yamamoto, Hiroyuki Kugoh, Hisao Ito, Mitsuo Oshimura, Yoshiyuki Fujiwara, Futoshi Okada
ABSTRACT
The human amphoterin-induced gene and open reading frame (AMIGO) was identified as a novel cell adhesion molecule of type I transmembrane protein. AMIGO2 is one of three members of the AMIGO family (AMIGO1, 2, and 3), and the similarity between them is approximately 40% at the amino acid level. We have previously shown that AMIGO2 functions as a driver of liver metastasis. Immunohistochemical analysis of AMIGO2 expression in colorectal cancer (CRC) using a commercially available anti-AMIGO2 mouse monoclonal antibody clone sc-373699 (sc mAb) correlated with liver metastasis and poor prognosis. However, the sc mAb was found to be cross-reactive with all three molecules in the AMIGO family. We generated a rat monoclonal antibody clone rTNK1A0012 (rTNK mAb) for human AMIGO2. The rTNK mAb was used to re-evaluate the association between AMIGO2 expression and liver metastases/clinical outcomes using the same CRC tissue samples previously reported with sc mAb. Western blot analysis revealed that a rTNK mAb was identified as being specific for AMIGO2 protein and did not cross-react with AMIGO1 and AMIGO3. The rTNK mAb and sc mAb showed higher AMIGO2 expression, which correlates with a high frequency of liver metastases (65.3% and 47.5%, respectively), while multivariate analysis showed that AMIGO2 expression was an independent prognostic factor for liver metastases (p = 7.930E-10 and p = 1.707E-5). The Kaplan-Meier analyses showed that the rTNK mAb (p = 0.004), but not sc mAb (p = 0.107), predicted worse overall survival in patients with high AMIGO2 expression. The relationship between AMIGO2 expression and poor disease-specific survival showed a higher level of significance for rTNK mAb (p = 0.00004) compared to sc mAb (p = 0.001). These results indicate that the developed rTNK1A0012 mAb is an antibody that specifically recognizes AMIGO2 by immunohistochemistry and can be a more reliable and applicable method for the diagnostic detection of liver metastases and worse prognosis in patients with high AMIGO2-expressing CRC.
Sigma-Aldrich
Rosetta-gami B(DE3)pLysS Competent Cells - Novagen, Rosetta-gami B strains combines the key features of BL21, Origami, and Rosetta to enhances the expression of eukaryotic proteins. T7 lysozyme expression suppresses basal T7 expression.
Sigma-Aldrich
Anti-AMIGO2 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Fetal Bovine Serum, sterile-filtered, non-USA origin, suitable for cell culture
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture